MilliporeSigma
  • In vivo and in vitro promutagen activation by Vicia faba of thiocarbamate herbicides molinate and butylate to products inducing sister chromatid exchanges in human lymphocyte cultures.
In vivo and in vitro promutagen activation by Vicia faba of thiocarbamate herbicides molinate and butylate to products inducing sister chromatid exchanges in human lymphocyte cultures.
Mutation research (1999-02-26)
M E Calderón-Segura, S Gómez-Arroyo, R Villalobos-Pietrini, M Espinosa-Ramírez
ABSTRACT
Molinate and butylate treatments for 4 h of Vicia faba root tip meristems, showed that both thiocarbamate herbicides increased significantly SCE frequency. Direct treatments of molinate and butylate on human lymphocytes applied 24 h after the beginning of culture did not induce SCE. When S10 extracts of the Vicia roots, treated for 4 h with molinate and butylate (in vivo activation) were added to lymphocytes (24 h after of the beginning of culture), SCE were induced in a concentration-response manner. The in vitro assays, in which molinate and butylate was added at 48 h lymphocyte cultures for 4 h, showed a negative response, however, in the treatment where the S10 metabolic mix was added the SCE frequencies were significantly different to the control, and the concentration-response relationship was not observed with molinate, but it was obtained with butylate. The results showed that both herbicides needed the V. faba metabolism to produce SCE in human lymphocyte culture.
Product Description
Supelco
Butylat, PESTANAL®, analytical standard