MilliporeSigma
  • Association between regulator of G protein signaling 9-2 and body weight.
Association between regulator of G protein signaling 9-2 and body weight.
PloS one (2011-12-02)
Jeffrey L Waugh, Jeremy Celver, Meenakshi Sharma, Robert L Dufresne, Dimitra Terzi, S Craig Risch, William G Fairbrother, Rachael L Neve, John P Kane, Mary J Malloy, Clive R Pullinger, Harvest F Gu, Christos Tsatsanis, Steven P Hamilton, Stephen J Gold, Venetia Zachariou, Abraham Kovoor
ABSTRACT
Regulator of G protein signaling 9-2 (RGS9-2) is a protein that is highly enriched in the striatum, a brain region that mediates motivation, movement and reward responses. We identified a naturally occurring 5 nucleotide deletion polymorphism in the human RGS9 gene and found that the mean body mass index (BMI) of individuals with the deletion was significantly higher than those without. A splicing reporter minigene assay demonstrated that the deletion had the potential to significantly decrease the levels of correctly spliced RGS9 gene product. We measured the weights of rats after virally transduced overexpression of RGS9-2 or the structurally related RGS proteins, RGS7, or RGS11, in the nucleus accumbens (NAc) and observed a reduction in body weight after overexpression of RGS9-2 but not RGS7 or 11. Conversely, we found that the RGS9 knockout mice were heavier than their wild-type littermates and had significantly higher percentages of abdominal fat. The constituent adipocytes were found to have a mean cross-sectional area that was more than double that of corresponding cells from wild-type mice. However, food intake and locomotion were not significantly different between the two strains. These studies with humans, rats and mice implicate RGS9-2 as a factor in regulating body weight.
Sigma-Aldrich
SIGMAFAST Protease Inhibitor Cocktail Tablets, EDTA-Free, for use in purification of Histidine-tagged proteins
Pricing and availability is not currently available.