MilliporeSigma
  • Eplerenone and chronic heart failure. No comparison with spironolactone.
Eplerenone and chronic heart failure. No comparison with spironolactone.
Prescrire international (2013-07-20)
ABSTRACT
Still no randomised controlled trials versus spironolactone, but a prematurely terminated placebo-controlled trial with 2737 patients.
Sigma-Aldrich
Spironolactone, 97.0-103.0%
Pricing and availability is not currently available.
Spironolactone, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.