MilliporeSigma
  • William H. Daughaday and the foundations of modern research into growth hormone and the insulin-like growth factors.
William H. Daughaday and the foundations of modern research into growth hormone and the insulin-like growth factors.
Pediatric endocrinology reviews : PER (2013-06-04)
Peter Rotwein
ABSTRACT
This vignette summarizes some of the scientific accomplishments of Dr. William H. Daughaday, a founder of modern research into the biological effects of growth hormone and the insulin-like growth factors, and formulator of the somatomedin hypothesis of GH actions on growth.