MilliporeSigma
  • Biosynthesis and characterization of a new copolymer, poly(3-hydroxyvalerate-co-5-hydroxydecenoate), from Pseudomonas aeruginosa.
Biosynthesis and characterization of a new copolymer, poly(3-hydroxyvalerate-co-5-hydroxydecenoate), from Pseudomonas aeruginosa.
Biotechnology letters (2012-12-25)
Pinkee Phukon, Binod Pokhrel, B K Konwar, S K Dolui
ABSTRACT
Using a new isolate of Pseudomonas aeruginosa, we obtained 7 g cell dry wt (CDW/l) using 5 % (w/v) glucose. Crude polyhydroxyalkanoates were obtained at 14.6 % of CDW. FTIR and NMR analysis confirmed that this was a new co-polymer: 3-hydroxyvalerate-co-5-hydroxydecenoate. Differential scanning calorimetry analysis showed two different melting temperatures of the copolymer and also indicated the glass transition temperature to be 4 °C. The polydispersity index of the polymer was 1.059.
Sigma-Aldrich
5-Hydroxydecanoic acid sodium salt, ≥97% (HPLC)