MilliporeSigma
  • Artificial electron carriers for photoenzymatic synthesis under visible light.
Artificial electron carriers for photoenzymatic synthesis under visible light.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-04-11)
Sahng Ha Lee, Hye Jung Lee, Keehoon Won, Chan Beum Park
ABSTRACT
NAD analogues can be employed as artificial electron carriers for photoenzymatic synthesis under visible light. Four different NAD analogues that have a 3-substituted pyridine ring have been investigated. 3-Acetylpyridine adenine dinucleotide and 3-pyridinealdehyde adenine dinucleotide were photochemically reduced much more efficiently than NAD, while their reduced products showed coenzyme activity comparable to natural NAD.
Sigma-Aldrich
3-Acetylpyridine adenine dinucleotide, ≥85%