MilliporeSigma
  • Palladium-catalyzed S-benzylation of unprotected mercaptobenzoic acid with benzyl alcohols in water.
Palladium-catalyzed S-benzylation of unprotected mercaptobenzoic acid with benzyl alcohols in water.
Organic & biomolecular chemistry (2012-03-10)
Hidemasa Hikawa, Yuusaku Yokoyama
ABSTRACT
Palladium-catalyzed S-benzylation of unprotected mercaptobenzoic acids with benzyl alcohols gave only S-benzylated mercaptobenzoic acids in good yields. Water may play an important role for the smooth generation of the (η(3)-benzyl)palladium species by activation of the hydroxyl group of the benzyl alcohol.
Sigma-Aldrich
4-Mercaptobenzoic acid, technical grade, 90%
Sigma-Aldrich
4-Mercaptobenzoic acid, 99%