MilliporeSigma
  • Sorption-desorption behavior of metsulfuron-methyl and sulfosulfuron in soils.
Sorption-desorption behavior of metsulfuron-methyl and sulfosulfuron in soils.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2012-03-02)
Neera Singh, Shashi Bala Singh
ABSTRACT
Sorption of metsulfuron-methyl and sulfosulfuron were studied in five Indian soils using batch sorption method. Freundlich adsorption equation described the sorption of herbicides with K(f) (adsorption coefficient) values ranging between 0.21 and 1.88 (metsulfuron-methyl) and 0.37 and 1.17 (sulfosulfuron). Adsorption isotherms were L-type suggesting that the herbicides sorption decreased with increase in the initial concentration of the herbicide in the solution. The K(f) for metsulfuron-methyl showed good positive correlation with silt content (significant at p = 0.01) and strong negative correlation with the soil pH (significant at p = 0.05) while sorption of sulfosulfuron did not correlate with any of the soil parameter. Desorption of herbicides was concentration dependent and, in general, sulfosulfuron showed higher desorption than the metsulfuron-methyl. The study indicates that these herbicides are poorly sorbed in the Indian soil types and there may be a possibility of their leaching to lower soil profiles.
Product Description
Supelco
Metsulfuron-methyl, PESTANAL®, analytical standard