MilliporeSigma
  • Zirconium phosphate nano-platelets: a novel platform for drug delivery in cancer therapy.
Zirconium phosphate nano-platelets: a novel platform for drug delivery in cancer therapy.
Chemical communications (Cambridge, England) (2012-01-06)
Agustín Díaz, Vipin Saxena, Julissa González, Amanda David, Barbara Casañas, Carrie Carpenter, James D Batteas, Jorge L Colón, Abraham Clearfield, M Delwar Hussain
ABSTRACT
Doxorubicin was intercalated into novel zirconium phosphate nano-platelets (ZrP). The obtained doxorubicin intercalated ZrP nano-platelets had an impressive 34.9% (w/w) drug loading. We used this material to deliver doxorubicin to breast cancer cells (MCF-7). Cellular studies with MCF-7 cells showed higher uptake and cytotoxicity of doxorubicin loaded ZrP compared to free doxorubicin.
Sigma-Aldrich
Zirconium(IV) hydrogenphosphate