MilliporeSigma
  • Preclinical testing of the Akt inhibitor triciribine in T-cell acute lymphoblastic leukemia.
Preclinical testing of the Akt inhibitor triciribine in T-cell acute lymphoblastic leukemia.
Journal of cellular physiology (2010-09-22)
Camilla Evangelisti, Francesca Ricci, Pierluigi Tazzari, Francesca Chiarini, Michela Battistelli, Elisabetta Falcieri, Andrea Ognibene, Pasqualepaolo Pagliaro, Lucio Cocco, James A McCubrey, Alberto M Martelli
ABSTRACT
Over the past 20 years, survival rates of T-cell acute lymphoblastic leukemia (T-ALL) patients have improved, mainly because of advances in polychemotherapy protocols. Despite these improvements, we still need novel and less toxic treatment strategies targeting aberrantly activated signaling networks which increase proliferation, survival, and drug resistance of T-ALL cells. One such network is represented by the phosphatidylinositol 3-kinase (PI3K)/Akt axis. PI3K inhibitors have displayed some promising effects in preclinical models of T-ALL. Here, we have analyzed the therapeutic potential of the Akt inhibitor, triciribine, in T-ALL cell lines. Triciribine caused cell cycle arrest and caspase-dependent apoptosis. Western blots demonstrated a dose-dependent dephosphorylation of Akt1/Akt2, and of mammalian target of rapamycin complex 1 downstream targets in response to triciribine. Triciribine induced autophagy, which could be interpreted as a defensive mechanism, because an autophagy inhibitor (chloroquine) increased triciribine-induced apoptosis. Triciribine synergized with vincristine, a chemotherapeutic drug employed for treating T-ALL patients, and targeted the side population of T-ALL cell lines, which might correspond to leukemia initiating cells. Our findings indicate that Akt inhibition, either alone or in combination with chemotherapeutic drugs, may serve as an efficient treatment towards T-ALL cells requiring upregulation of this signaling pathway for their proliferation and survival.
Sigma-Aldrich
Triciribine hydrate, ≥97% (HPLC)
Pricing and availability is not currently available.