MilliporeSigma
[Corneal tatoo--art or science?].
Oftalmologia (Bucharest, Romania : 1990) (2009-08-25)
Andreea-Madalina Craiu
ABSTRACT
The permanent colouring of disfigured corneal scars is known for almost 200 years. Because of improvement in surgical reconstructive techniques, corneal tattoing is used today only with a restricted group on carefully chosen patients, and merely for esthetique reasons.