MilliporeSigma
  • [Determination of major elements in soil from cancer village by X-ray fluorescence spectrometry].
[Determination of major elements in soil from cancer village by X-ray fluorescence spectrometry].
Guang pu xue yu guang pu fen xi = Guang pu (2009-03-11)
Zhen-Lin Wei, He Li, Yu-Kui Rui
ABSTRACT
Many social problems arise from environmental pollution, cancer village is one of the many important problems caused by pollution. The authors selected a typical cancer village where 80-100 people died of cancer in the last five years, but there are only a total of 1 200 people in this village. The authors sampled soils from crops-planted areas and detected the major elements by X-ray fluorescence spectrometry. The results showed that the contents of SiO2, TiO2, Al2O3, Fe2O3, MnO, MgO, CaO, Na2O, K2O and P2O5 in soil of this village were 66.05%, 0.66%, 11.37%, 3.93%, 0.075%, 1.97%, 5.47%, 1.90%, 2.11% and 0.20% respectively; with the precision being +/- 0.20%, +/- 0.005%, +/- 0.10%, +/- 0.10%, +/- 0.005%, +/- 0.05%, +/- 0.04%, +/- 0.08%, +/- 0.02% and +/- 0.005% respectively, which showed that X-ray fluorescence spectrometry is a good method.
Sigma-Aldrich
Sodium oxide, 80%