MilliporeSigma
  • Supramolecular synthesis of selenazoles using selenourea in water in the presence of beta-cyclodextrin under atmospheric pressure.
Supramolecular synthesis of selenazoles using selenourea in water in the presence of beta-cyclodextrin under atmospheric pressure.
The Journal of organic chemistry (2007-02-03)
M Narender, M Somi Reddy, V Pavan Kumar, V Prakash Reddy, Y V D Nageswar, K Rama Rao
ABSTRACT
Selenazoles were synthesized from alpha-bromo ketones and selenourea in the presence of beta-cyclodextrin in water at 50 degrees C under atmospheric pressure.
Sigma-Aldrich
Selenourea, 98%