MilliporeSigma
  • Rapid access to uronic acid-based mimetics of Kdn2en from D-glucurono-6,3-lactone.
Rapid access to uronic acid-based mimetics of Kdn2en from D-glucurono-6,3-lactone.
Carbohydrate research (2000-11-28)
P Florio, R J Thomson, M von Itzstein
ABSTRACT
A concise route to novel mimetics of Kdn2en, based on delta4-uronic acids, from D-glucurono-6,3-lactone is presented. Uronic acid-based mimetics in which an aliphatic ether (O-glycoside), a thioether (S-glycoside), or acetamide takes the place of the natural C-6 glycerol sidechain of the sialic acid were synthesized from the key intermediate, methyl 2,3,4-tri-O-acetyl-alpha-D-glucopyranosyluronate bromide.
Sigma-Aldrich
D-(+)-Glucuronic acid γ-lactone, ≥99%
Pricing and availability is not currently available.