MilliporeSigma
Fate and transport of ethoprophos in the Jamaican environment.
The Science of the total environment (1999-11-24)
D E Robinson, A Mansingh, T P Dasgupta
ABSTRACT
The hydrolytic half lives of ethoprophos in distilled, river, brackish and open sea water were 25, 133, 65 and 81 days, respectively. Under laboratory conditions, volatilisation of the residues after 12 h was 1.4-3.6, 2.3-4.5 and 6.5-20.2% from a sandy loam soil with 1, 10 and 20% moisture levels, respectively. Photolysis in soil was significantly faster (P < 0.05) in direct sunlight (T1/2 of 4.7 days) than in the shade (T1/2 of 12.3 days). The microbial degradation of ethoprophos was more than two-fold faster in unsterile soil (T1/2 of 10.9 days) than in sterile soil (T1/2 of 28.8 days). The runoff of ethoprophos from unweeded plantation soil at 23 degrees slope was significantly (P = 0.015) less than at 38 degrees slope; the amounts lost after 9 weeks and 27.5 mm of rainfall were 89.4 and 91.2%, respectively, of the applied amount from the two respective slopes. In the weeded plots, 93.6 and 92.4% of the applied insecticide were lost from 23 degrees and 38 degrees slopes, respectively. Under laboratory conditions, between 67.0 and 85.1% of ethoprophos leached through the soil columns. Under field conditions, after 9 weeks and 25 mm of rainfall, only 2.8 and 2.0% residues were recovered at a depth of 10-15 cm from unweeded and weeded slopes, respectively at 23 degrees slope, and 2.2 and 1.9% from the two respective plots at 38 degrees slope.
Product Description
Supelco
Ethoprophos, PESTANAL®, analytical standard