MilliporeSigma
  • Application of two-dimensional mapping for an analysis of galactosyllactoses in yogurt.
Application of two-dimensional mapping for an analysis of galactosyllactoses in yogurt.
Bioscience, biotechnology, and biochemistry (1998-08-28)
T Saito, T Tsuji, H Kitazawa, Y Kawai, T Itoh
ABSTRACT
A two-dimensional mapping analysis was performed by HPLC for 4 kinds of standard galactosyllactoses (GLs, trisaccharide) which were assumed to be produced from lactose (galactopyranosyl beta 1-->4 glucopyranose) in yogurt during the fermentation of lactic acid bacteria. After the pyridylamination of GLs, they were analyzed by HPLC in the reverse-phase (RP) and anion-exchange (AE) modes. The retention times of each peak obtained were converted to glucose units (GU) in RP mode for the pyridylaminated isomaltooligosaccharides (G1-3) and to relative retention time (RRT) in AE mode against pyridylaminated-isomaltotriose, and then the address data [GU, RRT] were plotted on a graph. This two-dimensional mapping method was found useful for a rapid qualitative evaluation of the chemical structure of trisaccharides formed in yogurt.