MilliporeSigma
Synthesis of 3'-azolyl-2',3'-dideoxyhexose nucleosides.
Acta chemica Scandinavica (Copenhagen, Denmark : 1989) (1998-07-14)
K Walczak, E B Pedersen, C Nielsen
ABSTRACT
1,8-Diazabicyclo[5.4.0]undec-7-ene salts of 2-methyl-4(5)-nitroimidazole or benzotriazole were obtained in crystalline form. Michael-type addition of these salts to (4S,5R)-(E)-4,6-di-O-acetyl-5-hydroxy-2-hexenal gave, after acetylation of the product, an isomeric mixture of acetylated 3-(azol-1-yl)-2,3-dideoxy-D-arabino-hexopyranosides and 3-(azol-1-yl)-2,3-dideoxy-D-ribo-hexofuranosides. Reaction of these peracetylated adducts with trimethylsilylated thymine in the presence of trimethylsilyl trifluoromethanesulfonate (TMS triflate) afforded the corresponding nucleosides which were deprotected by using methanolic ammonia. The nucleosides were found inactive against HIV-1 and HSV-1.
Product Description
Supelco
Tinidazole Related Compound A, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Methyl-4(5)-nitroimidazole, 99%
Metronidazole impurity A, European Pharmacopoeia (EP) Reference Standard
Supelco
Menidazole, VETRANAL®, analytical standard