MilliporeSigma
  • Prenylation of olivetolate by a hemp transferase yields cannabigerolic acid, the precursor of tetrahydrocannabinol.
Prenylation of olivetolate by a hemp transferase yields cannabigerolic acid, the precursor of tetrahydrocannabinol.
FEBS letters (1998-06-02)
M Fellermeier, M H Zenk
ABSTRACT
A new enzyme, geranylpyrophosphate:olivetolate geranyltransferase (GOT), the first enzyme in the biosynthesis of cannabinoids could be detected in extracts of young leaves of Cannabis sativa. The enzyme accepts geranylpyrophosphate (GPP) and to a lesser degree also nerylpyrophosphate (NPP) as a cosubstrate. It is, however, specific for olivetolic acid; its decarboxylation product olivetol is inactive as a prenyl acceptor.
Sigma-Aldrich
Olivetol, 95%