MilliporeSigma
  • Transformation of nitrobenzene by ferredoxin NADP oxidoreductase from spinach leaves.
Transformation of nitrobenzene by ferredoxin NADP oxidoreductase from spinach leaves.
Biochemical and biophysical research communications (1998-01-22)
M M Shah, J A Campbell
ABSTRACT
Nitrobenzene was reduced in a solution containing ferredoxin NADP oxidoreductase (FNR) from spinach leaves and NADPH generating system. The product of nitrobenzene was identified as phenylhydroxylamine (PHA) on 1:1 basis.
Sigma-Aldrich
N-Phenylhydroxylamine, ≥95.0%
Pricing and availability is not currently available.