MilliporeSigma
  • Sample preparation for taxol and cephalomannine determination in various organs of Taxus sp.
Sample preparation for taxol and cephalomannine determination in various organs of Taxus sp.
Journal of pharmaceutical and biomedical analysis (1996-06-01)
K Glowniak, G Zgórka, A Józefczyk, M Furmanowa
ABSTRACT
Solid-phase extraction and preparative thin-layer chromatography were applied as sample preparation techniques for the purification of crude extracts from twigs and needles of various Taxus species as well as for the isolation of taxol and cephalomannine for further reversed phase high performance liquid chromatography analysis. Significant differences in the contents of taxanes examined were found. The preparative chromatographic methods used were compared and evaluated as routine and reproducible procedures for the rapid isolation and determination of taxol and cephalomannine in plant extracts.
Sigma-Aldrich
Cephalomannine, ≥97% (HPLC), solid