MilliporeSigma
  • Halopicolinic acids, novel products arising through the degradation of chloro- and bromo-biphenyl by Sphingomonas paucimobilis BPSI-3.
Halopicolinic acids, novel products arising through the degradation of chloro- and bromo-biphenyl by Sphingomonas paucimobilis BPSI-3.
Canadian journal of microbiology (1996-01-01)
A D Davison, P Karuso, D R Jardine, D A Veal
ABSTRACT
Sphingomonas paucimobilis BPSI-3 was previously isolated from a mixed microbial consortium growing on biphenyl as the sole source of carbon and energy. Transformation of 4-chlorobiphenyl (4CBP) was demonstrated by this strain, although little or no growth was observed. In minimal salts medium supplemented with 4CBP or bromobiphenyl and dextrose, yellow coloured product(s) were rapidly formed. Gas chromatography-mass spectrometry (GC-MS) revealed single-ring N-heterocyclic compounds that were identified as halopicolinic acids. We believe this to be the first report of such compounds being formed via biological transformation of halobiphenyls. A mechanism is proposed for their formation.
Sigma-Aldrich
4-Bromobiphenyl, 98%
Pricing and availability is not currently available.