MilliporeSigma
  • Suppression of intestinal smooth muscle contraction by 4-ethylguaiacol, a constituent of wood creosote.
Suppression of intestinal smooth muscle contraction by 4-ethylguaiacol, a constituent of wood creosote.
Pharmacology (1993-11-01)
M Toyoda, N Ogata, T Shibata
ABSTRACT
Wood creosote, a mixture of phenolic compounds, suppresses in vitro contractions of rat intestine. To identify a compound in wood creosote able to inhibit intestinal motility, we screened its constituent phenolic compounds and found 4-ethylguaiacol (4-EG) as an active compound. It suppressed the spontaneous phasic (IC50 = 513 +/- 48 mumol/l) as well as spasmogenic-agent-induced tonic longitudinal contractions of isolated rat ileum in a reversible and concentration-dependent manner. KCl-depolarization-induced tonic contraction, which was susceptible to a calcium channel blocking agent, was also suppressed by 4-EG with an IC50 of 433 +/- 41 mumol/l. Furthermore, calcium-ionophore-induced contraction, which was affected by an influx of extracellular calcium ion that bypassed calcium channels, was suppressed by 4-EG with an IC50 of 97 +/- 18 mumol/l. These results support the concept that the effect of wood creosote to suppress intestinal motility is attributable, partially or entirely, to its component 4-EG and that this effect of 4-EG on the intestinal muscle is produced at some stage(s) of the muscle contraction process after influx of extracellular calcium into the cytosol of smooth muscle.
Sigma-Aldrich
4-Ethylguaiacol, ≥98%, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
4-Ethylguaiacol, natural, FG
Pricing and availability is not currently available.