MilliporeSigma
Effect of fluconazole on theophylline disposition in humans.
European journal of clinical pharmacology (1994-01-01)
H Konishi, K Morita, A Yamaji
ABSTRACT
The effect of fluconazole, an antimycotic that inhibits cytochrome P-450-mediated drug metabolism, on theophylline kinetics and the production of its metabolites were compared with those of enoxacin in 5 healthy subjects. All subjects received a single oral dose of 240 mg theophylline (aminophylline, 300 mg) after they had been given oral fluconazole 100 mg every 12 h or enoxacin 200 mg every 8 h for three consecutive days. Pretreatment with enoxacin decreased the total clearance (CLT) and elimination rate constant (Kel) of theophylline by 50% and 46%, respectively, without changing the volume of distribution (Vd), but there were no significant change in any pharmacokinetic parameter when fluconazole was administered. Enoxacin led to a 50% reduction in the metabolic clearance (CLM) of theophylline and to decreases of 69%, 59% and 38% in the formation clearance of the three theophylline metabolites, 3-methylxanthine (3-MX), 1-methyluric acid (1-MU), and 1,3-dimethyluric acid (1,3-DMU), respectively, accompanied by significant changes in the urinary recovery of theophylline and its metabolites. In contrast, treatment with fluconazole led only to a slight decrease in the CLM of theophylline (16%) and in the formation clearance of its metabolites (15%-18%), and there was no change in the renal clearance (CLR) of theophylline. The results indicate that fluconazole is a minor inhibitor of theophylline disposition compared with enoxacin, and they suggest that the inhibitory action of fluconazole is selective for certain cytochrome P-450 isozymes, but not for the cytochrome P-4501A involved in theophylline metabolism.
Sigma-Aldrich
3-Methylxanthine, 98%