MilliporeSigma
  • Platelet-activating factor increases vascular resistance in rat hindquarters by thromboxane A2.
Platelet-activating factor increases vascular resistance in rat hindquarters by thromboxane A2.
The American journal of physiology (1995-05-01)
A Rizzo, A E Taylor, M I Townsley, P Logsdon, P L Khimenko, J R Fletcher
ABSTRACT
The effects of platelet-activating factor (PAF) on vascular resistance and capillary permeability were studied in the isolated rat hindquarter. Six groups were studied (n = 30): control; PAF alone (1.4 microM); and PAF (1.4 microM) pretreated with ibuprofen (30 mg/kg), thromboxane A2 (TxA2)-receptor antagonist (BM-13505, 2 mg/kg), PAF-receptor antagonist (WEB-2086, 5 mg/kg), or dexamethasone (5 mg/kg). The vascular resistance was calculated, and the reflection coefficient (sigma) was determined as an index of capillary permeability. Exogenous PAF caused a threefold increase in vascular resistance peaking at 5 min and a 2.5-fold increase in capillary permeability. The increased vascular resistance caused by PAF alone was significantly attenuated by ibuprofen, BM-13505, and dexamethasone. The PAF-induced permeability was neither attenuated by ibuprofen nor BM-13505. However, both the increased vascular resistance and permeability were blocked and attenuated by WEB-2086 and dexamethasone, respectively. We conclude that TxA2 mediates the PAF-induced increased vascular resistance; however, the increased vascular permeability is independent of the formation of TxA2 in the isolated hindquarter.
Sigma-Aldrich
Daltroban, >98% (HPLC)