MilliporeSigma
  • Assay of magnesium in serum and urine with use of only one enzyme, isocitrate dehydrogenase (NADP+).
Assay of magnesium in serum and urine with use of only one enzyme, isocitrate dehydrogenase (NADP+).
Clinical chemistry (1995-09-01)
T Fujita, Y Kawakami, S Kohda, S Takata, Y Sunahara, K Arisue
ABSTRACT
We report a method for assaying magnesium in serum and urine involving only one enzyme, isocitrate dehydrogenase (NADP+)(EC 1.1.1.42), which requires magnesium ion for activity. The enzymatic reduction of NADP+ by isocitrate increases in rate linearly up to at least 20 mmol/L magnesium in the presence of appropriate concentrations of the two metal-chelating reagents, EDTA and glycol ether diamine-N,N,N',N'-tetraacetate. Within-run (n = 20) CVs and day-to-day (n = 10) CVs for sera are < or = 1.5% and < or = 2.6%, respectively. Analytical recovery of magnesium in sera averages 96-100%. This method is not affected by bilirubin, hemoglobin, or lipemia. The method (y) gives the following results correlating with atomic absorption spectrophotometry (x): y = 1.03x + 0.06 mmol/L (n = 62, r = 0.995, Sylx = 0.03) for sera, and y = 1.03x - 0.10 mmol/L (n = 62, r = 0.989, Sylx = 0.19) for urines; with the calmagite method (x): y = 0.99x + 0.04 mmol/L (n = 62, r = 0.991, Sylx = 0.03) for sera, and y = 0.98x + 0.03 mmol/L (n = 62, r = 0.999, Sylx = 0.02) for urines.
Product Description
Supelco
Calmagite, indicator grade