MilliporeSigma
  • A single cytochrome P-450 system is involved in degradation of the herbicides EPTC (S-ethyl dipropylthiocarbamate) and atrazine by Rhodococcus sp. strain NI86/21.
A single cytochrome P-450 system is involved in degradation of the herbicides EPTC (S-ethyl dipropylthiocarbamate) and atrazine by Rhodococcus sp. strain NI86/21.
Applied and environmental microbiology (1995-05-01)
I Nagy, F Compernolle, K Ghys, J Vanderleyden, R De Mot
ABSTRACT
During atrazine degradation by Rhodococcus sp. strain N186/21, N-dealkylated metabolites and an hydroxyisopropyl derivative are produced. The cytochrome P-450 system that is involved in degradation of thiocarbamate herbicides by strain N186/21 (I. Nagy, G. Schoofs, F. Compernolle, P. Proost, J. Vanderleyden, and R. De Mot, J. Bacteriol. 177:676-687, 1995) is also required for atrazine degradation. Atrazine-degrading activity was conferred on the atrazine-negative strains, mutant FAJ2027 of Rhodococcus sp. strain N186/21 and Rhodococcus erythropolis SQ1, upon transformation with the genes encoding the cytochrome P-450 system.
Product Description
Supelco
EPTC, PESTANAL®, analytical standard