MilliporeSigma
  • Decay kinetics of N,N,N',N'-tetramethylbenzidine photocations in frozen micellar solution of cetyltrimethylammonium bromide.
Decay kinetics of N,N,N',N'-tetramethylbenzidine photocations in frozen micellar solution of cetyltrimethylammonium bromide.
Photochemistry and photobiology (1995-07-01)
T Wolff, S Weber, B Böhm, A Schütz
ABSTRACT
After UV irradiation of rapidly frozen solutions of N,N,N',N'-tetramethylbenzidine (TMB) in dilute aqueous micellar cetyltrimethylammonium bromide at 80 K, the ESR signal corresponding to the TMB+ cation was detected indicating photoionization. This signal was monitored as a function of time at various temperatures between 80 K and 260 K. On a timescale of a few days the signal decayed to a plateau value that was stable for months. Two distinct second-order processes for the decay from the initial to the plateau value were observed: one prevailing at low temperatures with an activation energy of 0.6 kJ/mol, and one prevailing at T > 220 K with an activation energy of 13 kJ/mol. By means of spin probe investigations the latter process was shown to be connected with a local softening of the region adjacent to the micellar surface.
Sigma-Aldrich
N,N,N′,N′-Tetramethylbenzidine, ≥95% (HPLC)
Pricing and availability is not currently available.