MilliporeSigma
  • Liposomes as immunological carriers for the preparation of antimannosyl antibodies.
Liposomes as immunological carriers for the preparation of antimannosyl antibodies.
Experientia (1982-05-15)
P K Das, P Ghosh, B K Bachhawat, M K Das
ABSTRACT
Antiserum was raised against an aminophenyl derivative of D-mannose grafted on to a liposomal surface. As characterized by immunodiffusion, quantitative precipitation and hapten inhibition, the antiserum was found to contain mannose specific antibodies in addition to antibodies against the aromatic phenyl group.
Sigma-Aldrich
4-Aminophenyl α-D-mannopyranoside, ≥98% (TLC)