MilliporeSigma
  • Binding of simple carbohydrates and some of their chromophoric derivatives to soybean agglutinin as followed by titrimetric procedures and stopped flow kinetics.
Binding of simple carbohydrates and some of their chromophoric derivatives to soybean agglutinin as followed by titrimetric procedures and stopped flow kinetics.
The Journal of biological chemistry (1984-06-10)
H De Boeck, H Lis, H van Tilbeurgh, N Sharon, F G Loontiens
ABSTRACT
The number of carbohydrate-binding sites of the GalNAc-specific lectin is four per tetramer. The binding parameters of N-acetyl-D-galactosamine and methyl-N-acetyl-alpha-D- galactosaminide , were determined by titrating the perturbation in the absorption spectrum of the protein. For D-galactosides, it was necessary to use p-nitrophenyl-N-acetyl-beta-D- galactosaminide as an indicator in substitution titrations. The association constants K were determined at several temperatures yielding 2.4 X 10(4) M-1 at 25 degrees C with delta H degree' = -45 kJ mol-1 and delta S degree' = -67 J X K-1 mol-1 for methyl-N-acetyl-alpha-D- galactosaminide and 1.0 X 10(3) M-1 at 25 degrees C, delta H degree' = -38 kJ mol-1 and delta S degree' = -69 J X K-1 mol-1 for methyl-alpha-D-galactoside. The increase in K by a factor of 25 caused by the acetamido group is largely enthalpic . Whenever different methods were used to determine the association constant of a given compound, the agreement was excellent. The observed changes in absorption or fluorescence of all chromophoric carbohydrate derivatives used are specific for the binding of carbohydrates. For large aromatic beta- aglycons such as p-nitrophenyl or 4-methylumbelliferyl groups, the increase in K of the N-acetyl-D- galactosaminide moiety is by a factor of 2 or less, but for a large N-5-dimethylaminonaphthalene-1-sulfonyl (dansyl) group this factor is about 20 as compared with the acetyl group. The concomitant 10-fold increase in dansyl fluorescence, also observed with four other GalNAc-binding lectins together with a favorable and large delta S degree' = +60 J X K-1 mol-1 strongly point at the presence of a hydrophobic region in the vicinity of the carbohydrate-binding site. The results of stopped flow kinetics with 4-methylumbelliferyl-N-acetyl-beta-D- galactosaminide and the lectin are consistent with a simple mechanism for which k+ = 1.1 X 10(4) M-1 S-1 and k- = 0.4 S-1 at 25 degrees C. This k- is slower than for any monosaccharide-lectin complex reported so far.
Sigma-Aldrich
4-Methylumbelliferyl N-acetyl-β-D-galactosaminide, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
4-Nitrophenyl N-acetyl-β-D-galactosaminide, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
4-Nitrophenyl N-acetyl-α-D-galactosaminide, substrate for N-acetyl-α-D-galactosaminidase
Pricing and availability is not currently available.