MilliporeSigma
  • Group-directed modification of bacteriorhodopsin by arylisothiocyanates. Labeling, identification of the binding site and topology.
Group-directed modification of bacteriorhodopsin by arylisothiocyanates. Labeling, identification of the binding site and topology.
Journal of molecular biology (1984-02-15)
H Sigrist, P R Allegrini, K Stauffer, J Schaller, N G Abdulaev, E E Rickli, P Zahler
ABSTRACT
Group-directed hydrophobic modification of membrane-integrated protein segments by arylisothiocyanates is applied to bacteriorhodopsin. Labeling of purple membrane with phenylisothiocyanate and 4-N,N'-dimethylamino-azobenzene-4'-isothiocyanate results in covalent modification of a unique lysine epsilon-amino group of bacteriorhodopsin. Lysine residue 41, located in the amino-terminal chymotryptic fragment, has been identified as the arylisothiocyanate binding site by established sequencing techniques. The phenylisothiocyanate binding site is not accessible for the aqueously soluble analog p-sulfophenylisothiocyanate. Furthermore, the acid-induced bathochromic shift of the bound chromophore reagent is not observed following acidification of 4-N,N'-dimethylamino-azobenzene-4'-isothiocyanate-labeled purple membrane. The modification thus occurs in the hydrophobic membrane domain, providing further evidence for intramembraneous disposition of the modified protein segment. Light-induced proton translocation is preserved in reconstituted vesicles containing either phenylisothiocyanate-modified or 4-N,N'-dimethylamino-azobenzene-4'-isothiocyanate-modified bacteriorhodopsin.
Sigma-Aldrich
4-(4-Isothiocyanatophenylazo)-N,N-dimethylaniline, 97%