MilliporeSigma
  • Comparative effects of p-chloroamphetamine and 1-(p-chlorophenyl)piperazine on 5-hydroxyindole concentration in rat brain.
Comparative effects of p-chloroamphetamine and 1-(p-chlorophenyl)piperazine on 5-hydroxyindole concentration in rat brain.
Research communications in chemical pathology and pharmacology (1980-07-01)
R W Fuller, H D Snoddy
ABSTRACT
1-(p-Chlorophenyl)piperazine (PCPP), a structural analog of p-chloroamphetamine (PCA), increased serotonin and decreased 5-hydroxyindoleacetic acid (5-HIAA) concentration in rat brain at 6 hours. At one day and one week, serotonin and 5-HIAA concentrations were normal. In contrast, PCA lowered serotonin and 5-HIAA at all three times. These different effects on 5-hydroxyindole concentrations in brain were not due to differences in brain levels of the two compounds, since these were approximately equal. PCPP lacks the ability of PCA to deplete brain 5-hydroxyindoles.
Sigma-Aldrich
1-(4-Chlorophenyl)piperazine, ≥98.0% (GC)