MilliporeSigma
  • Measurement of cytoplasmic, free magnesium concentration with entrapped eriochrome blue in nerve endings isolated from the guinea pig brain.
Measurement of cytoplasmic, free magnesium concentration with entrapped eriochrome blue in nerve endings isolated from the guinea pig brain.
Neuroscience letters (1986-12-03)
E Heinonen, K E Akerman
ABSTRACT
Cytoplasmic, free Mg2+ was measured spectrophotometrically using an intrasynaptosomally entrapped Mg2+-indicator, Eriochrome blue (EB). Addition of the ionophore A23187 or disruption of the synaptosomal plasma membrane with digitonin caused an increase in absorbance of entrapped EB with a maximum at 551 nm, which is typical for the Mg2+-EB complex. A conversion of absorbance changes to levels of free Mg2+ concentrations was performed after disruption of synaptosomal plasma membranes by digitonin. The results indicated that the internal, free Mg2+ increased from 0.34 to 2.2 mM when the extracellular Mg2+ concentration was increased from 1 to 5 mM. The low values of cytoplasmic, free Mg2+ concentrations suggest the presence of effective regulatory mechanisms in the nerve endings.