MilliporeSigma
  • Chemical modification and inactivation of rat liver microsomal cytochrome P-450c by 2-bromo-4'-nitroacetophenone.
Chemical modification and inactivation of rat liver microsomal cytochrome P-450c by 2-bromo-4'-nitroacetophenone.
The Journal of biological chemistry (1986-09-05)
A Parkinson, D E Ryan, P E Thomas, D M Jerina, J M Sayer, P J van Bladeren, M Haniu, J E Shively, W Levin
ABSTRACT
The alkylating agent 2-bromo-4'-nitroacetophenone (BrNAP) binds covalently to each of 10 isozymes of purified rat liver microsomal cytochrome P-450 (P-450a-P-450j) but substantially inhibits the catalytic activity of only cytochrome P-450c. Regardless of pH, incubation time, presence of detergents, or concentration of BrNAP, treatment of cytochrome P-450c with BrNAP resulted in no more than 90% inhibition of catalytic activity. Alkylation with BrNAP did not cause the release of heme from the holoenzyme or alter the spectral properties of cytochrome P-450c, data that exclude the putative heme-binding cysteine, Cys-460, as the major site of alkylation. Two residues in cytochrome P-450c reacted rapidly with BrNAP, for which reason maximal loss of catalytic activity was invariably associated with the incorporation of approximately 1.5 mol of BrNAP/mol of cytochrome P-450c. Two major radio-labeled peptides were isolated from a tryptic digest of [14C]BrNAP-treated cytochrome P-450c by reverse-phase high performance liquid chromatography. The amino acid sequence of each peptide was determined by microsequence analysis, but the identification of the residues alkylated by BrNAP was complicated by the tendency of the adducts to decompose when subjected to automated Edman degradation. However, results of competitive binding experiments with the sulfhydryl reagent 4,4'-dithiodipyridine identified Cys-292 as the major site of alkylation and Cys-160 as the minor site of alkylation by BrNAP in cytochrome P-450c.
Sigma-Aldrich
2-Bromo-4′-nitroacetophenone, 95%