MilliporeSigma
  • Distinct presynaptic control of dopamine release in striosomal and matrix areas of the cat caudate nucleus.
Distinct presynaptic control of dopamine release in striosomal and matrix areas of the cat caudate nucleus.
Proceedings of the National Academy of Sciences of the United States of America (1989-11-01)
M L Kemel, M Desban, J Glowinski, C Gauchy
ABSTRACT
By use of a sensitive in vitro microsuperfusion method, the cholinergic prsynaptic control of dopamine release was investigated in a prominent striosome (areas poor in acetylcholinesterase activity) located within the core of cat caudate nucleus and also in adjacent matrix area. The spontaneous release of [3H]dopamine continuously synthesized from [3H]tyrosine in the matrix area was found to be twice that in the striosomal area; the spontaneous and potassium-evoked releases of [3H]dopamine were calcium-dependent in both compartments. With 10(-6) M tetrodotoxin, 5 x 10(-5) M acetylcholine stimulated [3H]dopamine release in both striosomal and matrix areas, effects completely antagonized by atropine (10(-6) M), thus showing the involvement of muscarinic receptors located on dopaminergic nerve terminals. Experiments without tetrodotoxin revealed a more complex regulation of dopamine release in the matrix: (i) In contrast to results seen in the striosome, acetylcholine induced only a transient stimulatory effect on matrix dopamine release. (ii) Although 10(-6) M atropine completely abolished the cholinergic stimulatory effect on [3H]dopamine release in striosomal area, delayed and prolonged stimulation of [3H]dopamine release was seen with atropine in the matrix. The latter effect was completely abolished by the nicotinic antagonist pempidine (10(-5) M). Therefore, in the matrix, in addition to its direct (tetrodotoxin-insensitive) facilitatory action on [3H]dopamine release, acetylcholine exerts two indirect (tetrodotoxin-sensitive) opposing effects: an inhibition and a stimulation of [3H]dopamine release mediated by muscarinic and nicotinic receptors, respectively.
Sigma-Aldrich
1,2,2,6,6-Pentamethylpiperidine, 97%