MilliporeSigma
  • [Hyperuricemia and acute kidney injury following kidney transplantation : a case report].
[Hyperuricemia and acute kidney injury following kidney transplantation : a case report].
Hinyokika kiyo. Acta urologica Japonica (2013-04-05)
Shigeaki Nakazawa, Ryoichi Imamura, Yoshiyuki Yamamoto, Takuji Hayashi, Go Tanigawa, Kazutoshi Fujita, Masahiro Hosomi, Seiji Yamaguchi
ABSTRACT
We report a case of hyperuricemia and acute kidney injury associated with mizoribine (MZR). A 15- year-old male with congenital renal hypoplasia underwent kidney transplantation. We used tacrolimus extended release (0.15 mg/kg/day), mizoribine (MZR) (12 mg/kg/day), prednisolone and basiliximab as immunosuppressants. On the 35th post operative day, he complained of acute right chest pain, right inguinal pain and dyspnea. Serum uric acid and creatinine were elevated. Accordingly, we changed MZR to mycophenolate mofetil, and added allopurinol and potassium citrate. Gradually, the symptoms disappeared and renal function was improved. In this case, prolonged MZR metabolism, hyperuricemia and progressive renal dysfunction may have formed a vicious cycle. In conclusion, monitoring of serum uric acid level is necessary, especially when using a high dose MZR.
Sigma-Aldrich
Mizoribine, ≥98% (TLC)