MilliporeSigma
  • Silica-coated and annealed CdS nanowires with enhanced photoluminescence.
Silica-coated and annealed CdS nanowires with enhanced photoluminescence.
Optics express (2013-03-14)
Shan Liang, Min Li, Jia-Hong Wang, Xiao-Li Liu, Zhong-Hua Hao, Li Zhou, Xue-Feng Yu, Qu-Quan Wang
ABSTRACT
The CdS/SiO(2) core/shell nanowires (NWs) with controlled shell thickness were successfully synthesized and subsequently heat-treated at 500 °C. The influences of silica shell coating and annealing processes on their optical properties have been investigated. Compared with original CdS NWs, the annealed CdS/SiO(2) NWs exhibited an enhanced band-edge emission with slowed photoluminescence lifetime, while the intensity of defect emission decreased. The results were ascribed to the surface passivation and recrystallization by shell coating and annealing. We believe our finding would help improving the optical properties of semiconductor NWs, and facilitate its applications in various realms, such as nanoscale emitter, sensor, and photoelectric device.
Sigma-Aldrich
Cadmium selenide, -325 Mesh particle size, 99.99% trace metals basis, electronic grade
Pricing and availability is not currently available.