MilliporeSigma
  • Chitosan-tripolyphosphate submicron particles as the carrier of entrapped rutin.
Chitosan-tripolyphosphate submicron particles as the carrier of entrapped rutin.
Food chemistry (2013-02-28)
K Konecsni, N H Low, M T Nickerson
ABSTRACT
Chitosan (CH)-tripolyphosphate (TPP) submicron particles were formed as carriers of entrapped rutin, and the release properties characterized using simulated gastric juices and fluids of the small intestine. Particle size, charge and entrapment efficiencies were investigated as a function of the CH:TPP molar ratio (2.0:1.0-5.0:1.0). Size was found to decrease from ~814 nm for the 2.0:1:0 mass ratio to ~528 nm for the ratios between 2.5:1.0 and 4.0:1.0, and then again to ~322 nm for the 5:0:1.0 mass ratio, whereas all particles carried a positive surface charge, increasing from +21 to +59 mV as the ratio increased from 2.0:1.0 to 5.0:1.0. The percent entrapment was found to rise from 3.68% to 57.6% as the ratios increased from 2.0:1:0 to 4.0:1:0, before reaching a plateau. Submicron particles (4.0:1.0 mass ratio only) were found to retain rutin in simulated gastric fluids, whereas in conditions which simulated fluids from the small intestine, only 20% of the entrapped rutin was released and 80% remained absorbed to the CH:TPP carriers. Such particles have applications for the delivery of phenolics in food and natural health products.
Sigma-Aldrich
Sodium triphosphate pentabasic, purum p.a., ≥98.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium tripolyphosphate, technical grade, 85%
Pricing and availability is not currently available.