MilliporeSigma
  • Bioaccumulation of PCDD/Fs, PCBs and PBDEs by earthworms in field soils of an E-waste dismantling area in China.
Bioaccumulation of PCDD/Fs, PCBs and PBDEs by earthworms in field soils of an E-waste dismantling area in China.
Environment international (2013-02-19)
Hongtao Shang, Pu Wang, Thanh Wang, Yawei Wang, Haidong Zhang, Jianjie Fu, Daiwei Ren, Weihai Chen, Qinghua Zhang, Guibin Jiang
ABSTRACT
A total of 60 paired samples of earthworm, corresponding soil and wormcast were collected to investigate the bioaccumulation tendency of polychlorinated dibenzo-p-dioxins/dibenzofurans (PCDD/Fs), polychlorinated biphenyls (PCBs) and polybrominated diphenyl ethers (PBDEs) in earthworms from a typical E-waste dismantling area in east China. Significant correlations were observed for the total concentrations among different matrix types except for PCDD/Fs in soil and earthworm. The bioaccumulation tendency showed some differences among the contaminants. Calculated biota-soil accumulation factors (BSAFs) indicated that PCBs and PBDEs had higher bioaccumulation potential compared to PCDD/Fs, which was somewhat different from laboratory studies. The plot of mean BSAFs versus log Kow values for PCBs and PBDEs was well fitted by a second-order polynomial with the maximum BSAF at approximately log Kow of 6.5. While for PCDD/Fs, only a slightly decreasing trend was observed with increasing log Kow. Composition analysis indicated that tetra-, penta- and hexa-halogenated homologs had higher bioaccumulation levels, indicating that medium-halogenated congeners with log Kow around 6.5 are more easily accumulated by earthworms. Furthermore, the ratios of BDE-47/-99 and BDE-99/-100 showed some discrepancies with the technical products and other biotic species, suggesting different bioaccumulation potential of PBDEs in earthworm.