MilliporeSigma
  • Catalytic signal amplification using [Fe(III)(biuret-amide)]-mesoporous silica nanoparticles: visual cyanide detection.
Catalytic signal amplification using [Fe(III)(biuret-amide)]-mesoporous silica nanoparticles: visual cyanide detection.
Chemical communications (Cambridge, England) (2013-02-09)
Chakadola Panda, Basab B Dhar, Bharmana Malvi, Yudhajit Bhattacharjee, Sayam Sen Gupta
ABSTRACT
Catalytic signal amplification was used for the colorimetric detection of CN(-) in aqueous media by using the enzyme catalase in tandem with mesoporous silica nanoparticle based synthetic HRP enzyme mimic Fe-MSNs. Signal amplification up to a maximum of eight fold was observed for the reporter "oxidized TMB" with respect to the added CN(-) ion.
Sigma-Aldrich
Biuret, 98% (Contains ≤10%(w/w) Triuret)