MilliporeSigma
  • Biomolecule-assisted construction of cadmium sulfide hollow spheres with structure-dependent photocatalytic activity.
Biomolecule-assisted construction of cadmium sulfide hollow spheres with structure-dependent photocatalytic activity.
Chemphyschem : a European journal of chemical physics and physical chemistry (2013-01-09)
Chengzhen Wei, Wenzhe Zang, Jingzhou Yin, Qingyi Lu, Qun Chen, Rongmei Liu, Feng Gao
ABSTRACT
In this study, we report the synthesis of monodispersive solid and hollow CdS spheres with structure-dependent photocatalytic abilities for dye photodegradation. The monodispersive CdS nanospheres were constructed with the assistance of the soulcarboxymthyi chitosan biopolymer under hydrothermal conditions. The solid CdS spheres were corroded by ammonia to form hollow CdS nanospheres through a dissolution-reprecipitation mechanism. Their visible-light photocatalytic activities were investigated, and the results show that both the solid and the hollow CdS spheres have visible-light photocatalytic abilities for the photodegradation of dyes. The photocatalytic properties of the CdS spheres were demonstrated to be structure dependent. Although the nanoparticles comprising the hollow spheres have larger sizes than those comprising the solid spheres, the hollow CdS spheres have better photocatalytic performances than the solid CdS spheres, which can be attributed to the special hollow structure.