MilliporeSigma
  • Magnetic resonance imaging of soft tissue infection with iron oxide labeled granulocytes in a rat model.
Magnetic resonance imaging of soft tissue infection with iron oxide labeled granulocytes in a rat model.
PloS one (2012-12-14)
Hassina Baraki, Norman Zinne, Dirk Wedekind, Martin Meier, André Bleich, Silke Glage, Hans-Juergen Hedrich, Ingo Kutschka, Axel Haverich
ABSTRACT
We sought to detect an acute soft tissue infection in rats by magnetic resonance imaging (MRI) using granulocytes, previously labeled with superparamagnetic particles of iron oxide (SPIO). Parasternal infection was induced by subcutaneous inoculation of Staphylococcus aureus suspension in rats. Granulocytes isolated from isogenic donor rats were labeled with SPIO. Infected rats were imaged by MRI before, 6 and 12 hours after intravenous injection of SPIO-labeled or unlabeled granulocytes. MR findings were correlated with histological analysis by Prussian blue staining and with re-isolated SPIO-labeled granulocytes from the infectious area by magnetic cell separation. Susceptibility effects were present in infected sites on post-contrast T2*-weighted MR images in all animals of the experimental group. Regions of decreased signal intensity (SI) in MRI were detected at 6 hours after granulocyte administration and were more pronounced at 12 hours. SPIO-labeled granulocytes were identified by Prussian blue staining in the infected tissue and could be successfully re-isolated from the infected area by magnetic cell separation. The application of SPIO-labeled granulocytes in MRI offers new perspectives in diagnostic specificity and sensitifity to detect early infectious processes.
Sigma-Aldrich
Iron(III) ferrocyanide