MilliporeSigma
Azobenzene-based supramolecular polymers for processing MWCNTs.
Nanoscale (2012-12-12)
Laura Maggini, Tomas Marangoni, Benoit Georges, Joanna M Malicka, K Yoosaf, Andrea Minoia, Roberto Lazzaroni, Nicola Armaroli, Davide Bonifazi
ABSTRACT
Photothermally responsive supramolecular polymers containing azobenzene units have been synthesised and employed as dispersants for multi-walled carbon nanotubes (MWCNTs) in organic solvents. Upon triggering the trans-cis isomerisation of the supramolecular polymer intermolecular interactions between MWCNTs and the polymer are established, reversibly affecting the suspensions of the MWCNTs, either favouring it (by heating, i.e. cis→trans isomerisation) or inducing the CNTs' precipitation (upon irradiation, trans→cis isomerisation). Taking advantage of the chromophoric properties of the molecular subunits, the solubilisation/precipitation processes have been monitored by UV-Vis absorption spectroscopy. The structural properties of the resulting MWCNT-polymer hybrid materials have been thoroughly investigated via thermogravimetric analysis (TGA), X-ray photoelectron spectroscopy (XPS), transmission electron microscopy (TEM) and atomic force microscopy (AFM) and modelled with molecular dynamics simulations.
Sigma-Aldrich
Azobenzene, 98%
Pricing and availability is not currently available.