MilliporeSigma
  • Fingerprint analysis of Radix Aconiti using ultra-performance liquid chromatography-electrospray ionization/ tandem mass spectrometry (UPLC-ESI/MS n) combined with stoichiometry.
Fingerprint analysis of Radix Aconiti using ultra-performance liquid chromatography-electrospray ionization/ tandem mass spectrometry (UPLC-ESI/MS n) combined with stoichiometry.
Talanta (2012-12-04)
Hongbin Zhu, Chunyan Wang, Yao Qi, Fengrui Song, Zhiqiang Liu, Shuying Liu
ABSTRACT
A fingerprinting approach was developed by means of UPLC-ESI/MS(n) (ultra-performance liquid chromatography-electrospray ionization/mass spectrometry) for the quality control of processed Radix Aconiti, a widely used toxic traditional herbal medicine. The present fingerprinting approach was based on the two processing methods recorded in Chinese Pharmacopoeia for the purpose of reducing the toxicity and ensuring the clinical therapeutic efficacy. Similarity evaluation, hierarchical cluster analysis and principal component analysis were performed to evaluate the similarity and variation of the samples. The results showed that the well processed, unqualified processed and the raw Radix Aconiti could be clustered reasonably corresponding to the contents of their constituents. The loading plot shows that the main chemical markers having the most influence on the discrimination amongst the qualified and unqualified samples were mainly some monoester diterpenoid aconitines and diester diterpenoid aconitines. Finally, the UPLC-UV and UPLC-ESI/MS(n) characteristic fingerprints were established according to the well processed and purchased qualified samples. At the same time, a complementary quantification method of six Aconitine-type alkaloids was developed using UPLC-UV and UPLC-ESI/MS. The average recovery of the monoester diterpenoid aconitines was 95.4-99.1% and the average recovery of the diester diterpenoid aconitines was 103-112%. The proposed combined quantification method by UPLC-UV and UPLC-ESI/MS allows the samples analyzed in a wide concentration range. Therefore, the established fingerprinting approach in combination with chemometric analysis provides a flexible and reliable method for quality assessment of toxic herbal medicine.
Sigma-Aldrich
Aconitine, ≥95% (HPLC), crystalline