MilliporeSigma
Odorant-binding protein. Characterization of ligand binding.
The Journal of biological chemistry (1990-04-15)
J Pevsner, V Hou, A M Snowman, S H Snyder
ABSTRACT
We have characterized the odorant binding properties of purified bovine odorant-binding protein (OBP) using as a ligand [3H]3,7-dimethyloctan-1-ol ([3H]DMO). A broad variety of odorants, including terpenes, aldehydes, esters, and musks, bind to OBP with affinities of 0.2 to 100 microM. Odorant affinities for OBP correlate most closely with their stimulation of an odorant-sensitive adenylyl cyclase as well as hydrophobicity. We also measured the kinetics of binding for the ligands, [3H]DMO and 2-isobutyl-3-[3H]methoxypyrazine. Dissociation of both is markedly accelerated in the presence of excess unlabeled ligand. Competition curves of displacers for [3H]DMO binding are shallow, and saturation binding isotherms for 3H-odorants are curvilinear. These kinetic and equilibrium binding properties suggest that OBP interactions with odorant ligands are negatively cooperative.
Sigma-Aldrich
3,7-Dimethyl-1-octanol, ≥98%
Pricing and availability is not currently available.