MilliporeSigma
  • Electrochemical communication between microbial cells and electrodes via osmium redox systems.
Electrochemical communication between microbial cells and electrodes via osmium redox systems.
Biochemical Society transactions (2012-11-28)
Kamrul Hasan, Sunil A Patil, Dónal Leech, Cecilia Hägerhäll, Lo Gorton
ABSTRACT
Electrochemical communication between micro-organisms and electrodes is the integral and fundamental part of BESs (bioelectrochemical systems). The immobilization of bacterial cells on the electrode and ensuring efficient electron transfer to the electrode via a mediator are decisive features of mediated electrochemical biosensors. Notably, mediator-based systems are essential to extract electrons from the non-exoelectrogens, a major group of microbes in Nature. The advantage of using polymeric mediators over diffusible mediators led to the design of osmium redox polymers. Their successful use in enzyme-based biosensors and BFCs (biofuel cells) paved the way for exploring their use in microbial BESs. The present mini-review focuses on osmium-bound redox systems used to date in microbial BESs and their role in shuttling electrons from viable microbial cells to electrodes.
Sigma-Aldrich
Osmium, powder, 99.9% trace metals basis
Pricing and availability is not currently available.