MilliporeSigma
  • An investigation into the adsorption of thorium(IV) from aqueous solutions by a carboxylate-functionalised graft copolymer derived from titanium dioxide-densified cellulose.
An investigation into the adsorption of thorium(IV) from aqueous solutions by a carboxylate-functionalised graft copolymer derived from titanium dioxide-densified cellulose.
Journal of environmental radioactivity (2012-11-17)
T S Anirudhan, S S Sreekumari, S Jalajamony
ABSTRACT
The use of a carboxylate-functionalized graft copolymer (PGTDC-COOH) based on titanium dioxide-densified cellulose (TDC) for the removal and recovery of thorium(IV) [Th(IV)] from industrial wastewater is reported in this paper. Infrared spectroscopy and X-ray diffractometer were used to characterize the adsorbent material. Batch equilibrium experiments showed that the adsorbent exhibited 98.6 ± 3.1% adsorption from an initial concentration of 10 mg/L Th(IV) solution at pH 5.0. The sorption kinetics have been analysed by pseudo-first-order and pseudo-second-order kinetic models and the adsorption kinetics was described by pseudo-second-order model. The experimental data obeyed Langmuir isotherm and the maximum monolayer adsorption capacity for Th(IV) was also found to be 92.23 ± 2.4 mg/g at 30 °C. The desorption capacity of HCl concentrations of different strengths ranging from 0.001 to 0.1 M was also studied. Maximum desorption of 98.2 ± 2.8% for Th(IV) occurred with 0.1 M HCl. The adsorption efficiency towards Th(IV) ion removal was tested using artificial sea water. Almost complete removal was possible with 3.5 g of the adsorbent from 1 L of the sea water.
Sigma-Aldrich
Poly(methacrylic acid, sodium salt) solution, average Mw 4,000-6,000, 40 wt. % in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(methacrylic acid, sodium salt) solution, average Mn ~5,400, average Mw ~9,500 by GPC, 30 wt. % in H2O
Pricing and availability is not currently available.