MilliporeSigma
Recombinant laccase: II. Medical biosensor.
Critical reviews in eukaryotic gene expression (2012-11-13)
Nicola Luigi Bragazzi, Eugenia Pechkova, Dora Scudieri, Tercio Bezerra Correia Terencio, Manuela Adami, Claudio Nicolini
ABSTRACT
Langmuir-Blodgett (LB) technology was used to build a high-sensitivity enzyme-based biosensor for medical purposes. Recombinant fungal laccase from Rigidoporous lignosus, as previously described, was used to catalyze a widely used antidepressant in a micromolar range, namely, clomipramine. The topological properties of the laccase thin film were characterized via LB π-A isotherm and AFM (mean roughness 8.22 nm, compressibility coefficient 37.5 m/N). The sensitivity of the biosensor was investigated via UV spectroscopy, and linearity was found in the absorbance peak shift at 400 nm at drug concentration varying up to 20 uM. The enzyme kinetics was subsequently investigated with potentiometric and amperometric measurements, and we found electronic transfer of at least 1 electron, k(s) 0.57 s(-1), diffusion coefficient 3 × 10(-6) cm(2)/s, K(cat) 6825.92 min(-1), K(M) 4.1 uM, K(cat)/K(M) 2.8 × 10(7) mol(-1) s(-1), sensitivity of 440 nA/uM, maximum velocity 1706.48 nA/s, and response time less than 5 s. The amperometric and potentiometric measurements were repeated after a month, confirming the stability of the biosensor.
Sigma-Aldrich
Clomipramine hydrochloride, ≥98% (HPLC), powder
Supelco
Clomipramine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®