MilliporeSigma
Genotoxicity evaluation of Mequindox in different short-term tests.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2012-10-16)
Awais Ihsan, Xu Wang, Hong-Gong Tu, Wei Zhang, Meng-Hong Dai, De-Peng Peng, Yu-Lian Wang, Ling-Li Huang, Dong-Mei Chen, Shazia Mannan, Yan-Fei Tao, Zhen-Li Liu, Zong-Hui Yuan
ABSTRACT
Quinoxaline-1,4-dioxides (QdNOs) are the potent heterocyclic N-oxides with interesting biological properties such as antibacterial, anticandida, antitubercular, anticancer and antiprotozoal activities. Here, we tested and compared the mequindox (MEQ) for mutagenic abilities in a battery of different short term tests according to OECD guidelines. When compared with the controls, a strong mutagenicity of MEQ and carbadox (CBX) was observed with an approximate concentration-effect relationship in Salmonella reverse mutation test, chromosome aberration test, unscheduled DNA synthesis assay and HGPRT gene mutation test, in the absence and presence of S(9)-mix. In in vivo micronucleus test, CBX produced significant increase in the proportion of micronucleus formation than MEQ in mice bone marrow cells. From these results, we can conclude that MEQ had a strong genotoxic potential to mammalian cells in vitro as well as in vivo and its mutagenicity is slightly higher than CBX. Our results, for the 1st time, discuss the genotoxic potential of MEQ. These results not only confirm the earlier findings about CBX but also extend the knowledge and awareness about the genotoxic risk of QdNO derivatives.