MilliporeSigma
  • Brønsted acid-assisted intramolecular aminohydroxylation of N-alkenylsulfonamides under heavy metal-free conditions.
Brønsted acid-assisted intramolecular aminohydroxylation of N-alkenylsulfonamides under heavy metal-free conditions.
The Journal of organic chemistry (2012-10-12)
Katsuhiko Moriyama, Yuta Izumisawa, Hideo Togo
ABSTRACT
The intramolecular aminohydroxylation of N-alkenylsulfonamides proceeded under heavy metal-free conditions to give substituted prolinol derivatives in high yields. Oxone activated by catalytic Brønsted acid worked well as an electrophilic oxidant for this reaction.
Sigma-Aldrich
(S)-(+)-2-Pyrrolidinemethanol, 97%