MilliporeSigma
  • Rebamipide (OPC-12759) in the treatment of dry eye: a randomized, double-masked, multicenter, placebo-controlled phase II study.
Rebamipide (OPC-12759) in the treatment of dry eye: a randomized, double-masked, multicenter, placebo-controlled phase II study.
Ophthalmology (2012-09-27)
Shigeru Kinoshita, Saki Awamura, Kazuhide Oshiden, Norihiro Nakamichi, Hiroyuki Suzuki, Norihiko Yokoi
ABSTRACT
To investigate the dose response for efficacy of 1% and 2% rebamipide ophthalmic suspension compared with placebo in patients with dry eye. A randomized, double-masked, multicenter, placebo-controlled, parallel-group, dose-response phase II study. A total of 308 patients with dry eye. After a 2-week screening period, patients were randomized to receive placebo or 1% rebamipide or 2% rebamipide administered as 1 drop in each eye 4 times daily for 4 weeks. The primary objective end point was change in fluorescein corneal staining (FCS) score from baseline to last observation carried forward (LOCF). Secondary objective end points were lissamine green conjunctival staining (LGCS) score, tear film break-up time (TBUT), and the Schirmer's test. Secondary subjective end points included dry eye-related ocular symptoms (foreign body sensation, dryness, photophobia, eye pain, and blurred vision) score and patients' overall treatment impression score. Rebamipide dose response was observed in FCS, LGCS, and TBUT scores. Both 1% and 2% rebamipide were significantly more effective than the placebo in terms of the change from baseline to LOCF for FCS, LGCS, and TBUT scores. There was no significant difference between the rebamipide and placebo groups from baseline to LOCF in Schirmer's test values, and dose response was not observed. In the predefined dry eye subpopulation with a baseline FCS score of 10 to 15, the mean change from baseline in the 2% rebamipide group was larger than that in the 1% rebamipide group. Change from baseline to LOCF for all 5 dry eye-related ocular symptom scores and patients' overall treatment impression showed significant improvements in the 1% and 2% rebamipide groups compared with the placebo group, except for photophobia in the 1% rebamipide group. No deaths or drug-related serious adverse events occurred in any treatment group. The incidence of ocular abnormalities was similar across the rebamipide and placebo groups. Rebamipide was effective in treating both objective signs and subjective symptoms of dry eye and were well tolerated in this 4-week study. Although 1% and 2% rebamipide were both efficacious, 2% rebamipide may be more effective than 1% rebamipide in some measures. Proprietary or commercial disclosure may be found after the references.
Sigma-Aldrich
Lissamine Green B, Dye content 60 %
Supelco
Green S, analytical standard